missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-hydroxybenzonitrile, 95%, Thermo Scientific Chemicals

Catalog No. AAH3223103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3223103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3223103 Supplier Thermo Scientific Chemicals Supplier No. H3223103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126162-38-7 | Molecular Formula: C7H3F2NO | Molecular Weight: 155.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126162-38-7
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD00269596
InChI Key UIJJJWADIVZXNT-UHFFFAOYSA-N
PubChem CID 2736978
IUPAC Name 2,3-difluoro-4-hydroxybenzonitrile
SMILES OC1=C(F)C(F)=C(C=C1)C#N

Specifications

Melting Point 145°C to 149°C
Quantity 1 g
Synonym 4-cyano-2,3-difluorophenol, 2,3-difluoro-4-cyanophenol, 2,3-difluoro-4-cyano phenol, benzonitrile,2,3-difluoro-4-hydroxy, benzonitrile, 2,3-difluoro-4-hydroxy, 2,3-difluoro-4-hydroxybenzenecarbonitrile, pubchem2222, acmc-1c5t8, ksc514s9h, attercop-chm at134987
Formula Weight 155.1
Percent Purity 95%
Chemical Name or Material 2,3-Difluoro-4-hydroxybenzonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only