missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diethylpyrazine 98.0+%, TCI America™

Catalog No. D15815ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15815ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D15815ML Supplier TCI America Supplier No. D15815ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15707-24-1
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00006151
InChI Key GZXXANJCCWGCSV-UHFFFAOYSA-N
Synonym pyrazine, 2,3-diethyl, unii-ad31a42vy0, pyrazine, diethyl, fema no. 3136, 2,3-diethyl-pyrazine, shoyu pyrazine, pubchem8606, 2,3-diethyl pyrazine, acmc-1brc9, dsstox_cid_27101
PubChem CID 27458
IUPAC Name 2,3-diethylpyrazine
SMILES CCC1=NC=CN=C1CC

Specifications

Boiling Point 71°C
Quantity 5 mL
Formula Weight 136.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Diethylpyrazine
EINECSNumber (5)-0950
TSCA Yes