accessibility menu, dialog, popup

UserName

H3CNNCH3

CAS RN 15707-24-1

IUPAC Name: 2,3-diethylpyrazine
Synonyms: 2,3-diethylpyrazine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: GZXXANJCCWGCSV-UHFFFAOYSA-N
SMILES: CCC1=NC=CN=C1CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results