missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Didehydro-3'-deoxythymidine 98.0+%, TCI America™

Catalog No. D35801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35801G 1 g
D35805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35801G Supplier TCI America Supplier No. D35801G

Chemical Identifiers

CAS 3056-17-5
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.22
MDL Number MFCD00132921
InChI Key XNKLLVCARDGLGL-HGXVMFPFNA-N
Synonym stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine
PubChem CID 18283
ChEBI CHEBI:63581
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O

Specifications

Melting Point 166°C
Color White
Quantity 1 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-Didehydro-3′-deoxythymidine
RTECSNumber XP2075000
TSCA No