missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyridine, 99%

Catalog No. AC113720100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113720100 10 g Glass bottle
AC113720500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113720100 Supplier Thermo Scientific Chemicals Supplier No. 113720100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2402-77-9 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2402-77-9
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006229
InChI Key MAKFMOSBBNKPMS-UHFFFAOYSA-N
Synonym 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x
PubChem CID 16988
IUPAC Name 2,3-dichloropyridine
SMILES ClC1=CC=CN=C1Cl

Specifications

Melting Point 67.0°C to 70.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)Cl2
Quantity 10 g
Beilstein 20,II,152
Solubility Information Solubility in water: slightly soluble
Formula Weight 147.99
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dichloropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 219-281-8

RUO – Research Use Only