Learn More
2,3-Dichloropyridine, 99%
CAS: 2402-77-9 | C5H3Cl2N | 147.99 g/mol
$91.27 - $310.70
Chemical Identifiers
| CAS | 2402-77-9 |
|---|---|
| Molecular Formula | C5H3Cl2N |
| Molecular Weight (g/mol) | 147.99 |
| MDL Number | MFCD00006229 |
| InChI Key | MAKFMOSBBNKPMS-UHFFFAOYSA-N |
| Synonym | 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x |
| PubChem CID | 16988 |
| IUPAC Name | 2,3-dichloropyridine |
| SMILES | ClC1=CC=CN=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1224106
|
Thermo Scientific Chemicals
A1224106 |
5 g |
Each for $91.27
|
|
|||||
|
AAA1224114
|
Thermo Scientific Chemicals
A1224114 |
25 g |
Each for $310.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2402-77-9 | |
| 147.99 | |
| MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
| 16988 | |
| ClC1=CC=CN=C1Cl |
| C5H3Cl2N | |
| MFCD00006229 | |
| 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x | |
| 2,3-dichloropyridine |
Specifications
| 2402-77-9 | |
| Unpleasant | |
| MFCD00006229 | |
| UN2811 | |
| 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x | |
| ClC1=CC=CN=C1Cl | |
| 147.99 | |
| 147.99 | |
| 2,3-Dichloropyridine |
| 65°C to 69°C | |
| C5H3Cl2N | |
| 5 g | |
| 109811 | |
| MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
| 2,3-dichloropyridine | |
| 16988 | |
| 99% |
Safety and Handling
GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 219-281-8
RTECSNumber : US8050000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only