missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH6373306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6373306 5 g
AAH6373303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6373306 Supplier Thermo Scientific Chemicals Supplier No. H6373306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 184416-84-0
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD01861977
InChI Key VGKZZKKTZKRCPA-UHFFFAOYSA-N
Synonym 2,3-dichloroisonicotinic acid, dichloroisonicotinic acid, 4-carboxy-2,3-dichloropyridine, 2,3-dichloro-4-pyridinecarboxylic acid, 2,3-dichloropyridine-4-carboxylicacid, 2,3-dichloro pyridine-4-carboxylic acid, pubchem22306, acmc-1cbyj, 2,3-dichloroisonicotinicacid, 2,3-dichloro-isonicotinic acid
PubChem CID 2736083
IUPAC Name 2,3-dichloropyridine-4-carboxylic acid
SMILES OC(=O)C1=C(Cl)C(Cl)=NC=C1

Specifications

Melting Point 203°C to 207°C
Quantity 5 g
Formula Weight 192
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,3-Dichloropyridine-4-carboxylic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only