accessibility menu, dialog, popup

UserName

NOOHClCl

CAS RN 184416-84-0

IUPAC Name: 2,3-dichloropyridine-4-carboxylic acid
Synonyms: 2,3-dichloropyridine-4-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: VGKZZKKTZKRCPA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results