missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyrazine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 4858-85-9
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00040964
InChI Key MLCNOCRGSBCAGH-UHFFFAOYSA-N
Synonym pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
PubChem CID 78575
IUPAC Name 2,3-dichloropyrazine
SMILES ClC1=NC=CN=C1Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC377040050
SDS View Documents
Thermo Scientific Chemicals
377040050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC377040250
SDS View Documents
Thermo Scientific Chemicals
377040250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC377041000
SDS View Documents
Thermo Scientific Chemicals
377041000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4858-85-9
148.97
MLCNOCRGSBCAGH-UHFFFAOYSA-N
78575
ClC1=NC=CN=C1Cl
C4H2Cl2N2
MFCD00040964
pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
2,3-dichloropyrazine

Specifications

4858-85-9
100.0
105.0°C to 107.0°C (50.0 mmHg)
97.5% min. (GC)
C4H2Cl2N2
5g
pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
MLCNOCRGSBCAGH-UHFFFAOYSA-N
2,3-dichloropyrazine
78575
98%
97.5
1.4310g/mL
102°C
Glass bottle
MFCD00040964
1.431
Solubility in water: insoluble
ClC1=NC=CN=C1Cl
148.97
148.98
2, 3-Dichloropyrazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 225-460-1

SDS
Documents

Documents

RUO – Research Use Only