missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzylamine, 97%

Catalog No. p-7045761
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1188406 5 g
AAL1188414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1188406 Supplier Thermo Scientific Chemicals Supplier No. L1188406

CAS: 39226-95-4 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39226-95-4
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00047927
InChI Key JHBVZGONNIVXFJ-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzylamine, 2,3-dichlorophenyl methanamine, 2,3-dichloro-benzylamine, 2,3-dichlorobenzyl amine, chembl13165, 1-2,3-dichlorophenyl methanamine, benzenemethanamine, 2,3-dichloro, 2,3-dichlorophenyl methylamine, pubchem16713, acmc-1ae0r
PubChem CID 587625
IUPAC Name (2,3-dichlorophenyl)methanamine
SMILES C1=CC(=C(C(=C1)Cl)Cl)CN

Specifications

Density 1.321
Boiling Point 124°C to 126°C (15 mmHg)
Flash Point 105°C (221°F)
Refractive Index 1.582
Quantity 5 g
UN Number UN2735
Beilstein 2639296
Sensitivity Air Sensitive
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorobenzylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only