missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorobenzyl Chloride 98.0+%, TCI America™
$54.06 - $156.59
Chemical Identifiers
| CAS | 3290-01-5 |
|---|---|
| Molecular Formula | C7H5Cl3 |
| Molecular Weight (g/mol) | 195.47 |
| MDL Number | MFCD00035817 |
| InChI Key | AIPJZPPOFWCJRC-UHFFFAOYSA-N |
| Synonym | alpha,2,3-Trichlorotoluene |
| PubChem CID | 76790 |
| IUPAC Name | 1,2-dichloro-3-(chloromethyl)benzene |
| SMILES | C1=CC(=C(C(=C1)Cl)Cl)CCl |
Chemical Identifiers
| 3290-01-5 | |
| 195.47 | |
| AIPJZPPOFWCJRC-UHFFFAOYSA-N | |
| 76790 | |
| C1=CC(=C(C(=C1)Cl)Cl)CCl |
| C7H5Cl3 | |
| MFCD00035817 | |
| alpha,2,3-Trichlorotoluene | |
| 1,2-dichloro-3-(chloromethyl)benzene |
Specifications
| 3290-01-5 | |
| 255°C | |
| MFCD00035817 | |
| 3265 | |
| AIPJZPPOFWCJRC-UHFFFAOYSA-N | |
| 1,2-dichloro-3-(chloromethyl)benzene | |
| 76790 | |
| ≥98.0% (GC) | |
| 2,3-Dichlorobenzyl Chloride |
| Yellow | |
| C7H5Cl3 | |
| 5 g | |
| alpha,2,3-Trichlorotoluene | |
| C1=CC(=C(C(=C1)Cl)Cl)CCl | |
| 195.47 | |
| 195.47 | |
| Liquid |
Safety and Handling
TSCA : No