missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7127565
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29205G 5 g
D292025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29205G Supplier TCI America Supplier No. D29205G
Only null left

Chemical Identifiers

CAS 57915-78-3
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD03701317
InChI Key PVIGUZZDWGININ-UHFFFAOYSA-N
PubChem CID 2734602
IUPAC Name 1-(bromomethyl)-2,3-dichlorobenzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)CBr

Specifications

Color White-Yellow
Boiling Point 144°C
Quantity 5 g
UN Number 3261
Synonym 2,3-dichlorobenzyl bromide, 1-bromomethyl-2,3-dichlorobenzene, 2,3-dichlorobenzylbromide, alpha-bromo-2,3-dichlorotoluene, benzene,1-bromomethyl-2,3-dichloro, benzene, 1-bromomethyl-2,3-dichloro, acmc-209m1b
Formula Weight 239.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3-Dichlorobenzyl Bromide