missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzyl bromide, 99%

Catalog No. AC365580050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC365580050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC365580050 Supplier Thermo Scientific Chemicals Supplier No. 365580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57915-78-3 | C7H5BrCl2 | 239.921 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57915-78-3
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD03701317
InChI Key PVIGUZZDWGININ-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzyl bromide, 1-bromomethyl-2,3-dichlorobenzene, 2,3-dichlorobenzylbromide, alpha-bromo-2,3-dichlorotoluene, benzene,1-bromomethyl-2,3-dichloro, benzene, 1-bromomethyl-2,3-dichloro, acmc-209m1b
PubChem CID 2734602
IUPAC Name 1-(bromomethyl)-2,3-dichlorobenzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29°C to 32°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 239.93
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2, 3-Dichlorobenzyl bromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only