missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. D3206500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3206500G 500 g
D320625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3206500G Supplier TCI America Supplier No. D3206500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2905-60-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00035937
InChI Key YBONBWJSFMTXLE-UHFFFAOYSA-N
Synonym benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc
PubChem CID 17944
IUPAC Name 2,3-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC(Cl)=C1Cl

Specifications

Color White
Boiling Point 140°C
Quantity 500 g
UN Number 3261
Formula Weight 209.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 2,3-Dichlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
TSCA Yes