Learn More
2,3-Dichlorobenzoyl chloride, 98%
CAS: 2905-60-4 | C7H3Cl3O | 209.45 g/mol
$91.27 - $674.34
Chemical Identifiers
| CAS | 2905-60-4 |
|---|---|
| Molecular Formula | C7H3Cl3O |
| Molecular Weight (g/mol) | 209.45 |
| MDL Number | MFCD00035937 |
| InChI Key | YBONBWJSFMTXLE-UHFFFAOYSA-N |
| Synonym | benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc |
| PubChem CID | 17944 |
| IUPAC Name | 2,3-dichlorobenzoyl chloride |
| SMILES | ClC(=O)C1=CC=CC(Cl)=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0950606
|
Thermo Scientific Chemicals
L0950606 |
5 g |
Each for $91.27
|
|
|||||
|
AAL0950614
|
Thermo Scientific Chemicals
L0950614 |
25 g |
Each for $238.32
|
|
|||||
|
AAL0950622
|
Thermo Scientific Chemicals
L0950622 |
100 g |
Each for $674.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2905-60-4 | |
| 209.45 | |
| YBONBWJSFMTXLE-UHFFFAOYSA-N | |
| 17944 | |
| ClC(=O)C1=CC=CC(Cl)=C1Cl |
| C7H3Cl3O | |
| MFCD00035937 | |
| benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc | |
| 2,3-dichlorobenzoyl chloride |
Specifications
| 2905-60-4 | |
| 140°C (14 mmHg) | |
| Pungent | |
| MFCD00035937 | |
| UN3261 | |
| Moisture sensitive | |
| YBONBWJSFMTXLE-UHFFFAOYSA-N | |
| 2,3-dichlorobenzoyl chloride | |
| 17944 | |
| 98% |
| 28°C to 32°C | |
| 167°C (332°F) | |
| C7H3Cl3O | |
| 5 g | |
| 2575973 | |
| benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc | |
| ClC(=O)C1=CC=CC(Cl)=C1Cl | |
| 209.45 | |
| 209.46 | |
| 2,3-Dichlorobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-811-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only