missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoyl chloride, 98%

Catalog No. AAL0950606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0950606 5 g
AAL0950614 25 g
AAL0950622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0950606 Supplier Thermo Scientific Chemicals Supplier No. L0950606

CAS: 2905-60-4 | C7H3Cl3O | 209.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-60-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00035937
InChI Key YBONBWJSFMTXLE-UHFFFAOYSA-N
Synonym benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc
PubChem CID 17944
IUPAC Name 2,3-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 28°C to 32°C
Boiling Point 140°C (14 mmHg)
Flash Point 167°C (332°F)
Odor Pungent
Quantity 5 g
UN Number UN3261
Beilstein 2575973
Sensitivity Moisture sensitive
Formula Weight 209.46
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-811-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only