missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzonitrile, 98%

CAS: 6574-97-6 | C7H3Cl2N | 172.008 g/mol

$234.78 - $801.45

Chemical Identifiers

CAS 6574-97-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016372
InChI Key OHDYZVVLNPXKDX-UHFFFAOYSA-N
Synonym benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b
PubChem CID 736567
IUPAC Name 2,3-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)Cl)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1480906
View Documents
Thermo Scientific Chemicals
A1480906
5 g
Each for $234.78
Only null left
 
AAA1480914
View Documents
Thermo Scientific Chemicals
A1480914
25 g
Each for $801.45
Only null left
 
Description

Description

2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
It is miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

6574-97-6
172.008
OHDYZVVLNPXKDX-UHFFFAOYSA-N
736567
C1=CC(=C(C(=C1)Cl)Cl)C#N
C7H3Cl2N
MFCD00016372
benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b
2,3-dichlorobenzonitrile

Specifications

6574-97-6
114°C (6 mmHg)
MFCD00016372
UN3439
benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b
OHDYZVVLNPXKDX-UHFFFAOYSA-N
2,3-dichlorobenzonitrile
736567
98%
60°C to 65°C
C7H3Cl2N
5 g
2438753
It is miscible with water.
C1=CC(=C(C(=C1)Cl)Cl)C#N
172.008
172.01
2,3-Dichlorobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only