missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzonitrile, 98%

Catalog No. AAA1480906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1480906 5 g
AAA1480914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1480906 Supplier Thermo Scientific Chemicals Supplier No. A1480906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6574-97-6 | C7H3Cl2N | 172.008 g/mol

2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
It is miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 6574-97-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016372
InChI Key OHDYZVVLNPXKDX-UHFFFAOYSA-N
Synonym benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b
PubChem CID 736567
IUPAC Name 2,3-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)Cl)C#N

Specifications

Melting Point 60°C to 65°C
Boiling Point 114°C (6 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 2438753
Solubility Information It is miscible with water.
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only