missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoic Acid 98.0+%, TCI America™

Catalog No. D33195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33195G 5 g
D331925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33195G Supplier TCI America Supplier No. D33195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-45-3
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
InChI Key QAOJBHRZQQDFHA-UHFFFAOYSA-N
Synonym benzoic acid, 2,3-dichloro, 2,3-dichloro benzoic acid, unii-cgp39qv5wf, benzoic acid, dichloro, 2,3-dichorobenzoic acid, cgp39qv5wf, dichlorobenzoic acid, lamotrigine impurity e, pubchem8198, acmc-209kmq
PubChem CID 5765
IUPAC Name 2,3-dichlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C(=O)O

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorobenzoic Acid
EINECSNumber (3)-0062&(3)-1410&(3)-1424
RTECSNumber DG6475000
TSCA No