missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoic acid, 98%

Catalog No. AAA1052209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1052209 10 g
AAA1052218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1052209 Supplier Thermo Scientific Chemicals Supplier No. A1052209

CAS: 50-45-3 | C7H4Cl2O2 | 191.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-45-3
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00002413
InChI Key QAOJBHRZQQDFHA-UHFFFAOYSA-N
Synonym benzoic acid, 2,3-dichloro, 2,3-dichloro benzoic acid, unii-cgp39qv5wf, benzoic acid, dichloro, 2,3-dichorobenzoic acid, cgp39qv5wf, dichlorobenzoic acid, lamotrigine impurity e, pubchem8198, acmc-209kmq
PubChem CID 5765
IUPAC Name 2,3-dichlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C(=O)O

Specifications

Melting Point 166°C to 170°C
Quantity 10 g
Beilstein 1946217
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-039-5
RTECSNumber DG6475000
TSCA Yes

RUO – Research Use Only