missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloroanisole 97.0+%, TCI America™

Catalog No. D2306500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2306500G 500 g
D230625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2306500G Supplier TCI America Supplier No. D2306500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1984-59-4
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00000545
InChI Key HFEASCCDHUVYKU-UHFFFAOYSA-N
Synonym 2,3-dichloroanisole, benzene, 1,2-dichloro-3-methoxy, 2,3-dichloromethoxybenzene, 2,3-dichloranisol, dichloroanisole 2,3-, 1,2-dichloro-3-methoxy-benzene, dichloroanisol, 2,3-dichloro-anisol, pubchem3680, 2,3-dichloro anisole
PubChem CID 16126
IUPAC Name 1,2-dichloro-3-methoxybenzene
SMILES COC1=CC=CC(Cl)=C1Cl

Specifications

Quantity 500 g
Formula Weight 177.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3-Dichloroanisole