missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloroaniline 98.0+%, TCI America™
Supplier: TCI America D0320500G
Specifications
| 2,3-Dichloroaniline | |
| White | |
| MFCD00007657 | |
| 3442 | |
| BRPSAOUFIJSKOT-UHFFFAOYSA-N | |
| 2,3-dichloroaniline | |
| 11844 | |
| 162.01 | |
| Crystals |
| 608-27-5 | |
| C6H5Cl2N | |
| 500 g | |
| benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine | |
| C1=CC(=C(C(=C1)Cl)Cl)N | |
| 162.013 | |
| CHEBI:46636 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 608-27-5 | |
| 162.013 | |
| BRPSAOUFIJSKOT-UHFFFAOYSA-N | |
| 11844 | |
| 2,3-dichloroaniline |
| C6H5Cl2N | |
| MFCD00007657 | |
| benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine | |
| CHEBI:46636 | |
| C1=CC(=C(C(=C1)Cl)Cl)N |
Safety and Handling
EINECSNumber : (3)-0261
RTECSNumber : CX9862625
TSCA : Yes