missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloroaniline 98.0+%, TCI America™

Catalog No. p-7126141
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032025G 25 g
D0320500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D032025G Supplier TCI America Supplier No. D032025G
Only null left

Chemical Identifiers

CAS 608-27-5
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007657
InChI Key BRPSAOUFIJSKOT-UHFFFAOYSA-N
Synonym benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine
PubChem CID 11844
ChEBI CHEBI:46636
IUPAC Name 2,3-dichloroaniline
SMILES C1=CC(=C(C(=C1)Cl)Cl)N

Specifications

Color White
Quantity 25 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,3-Dichloroaniline
EINECSNumber (3)-0261
RTECSNumber CX9862625
TSCA Yes