missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-6-nitroquinoxaline 98.0+%, TCI America™

Catalog No. D47575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47575G 5 g
D47571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47575G Supplier TCI America Supplier No. D47575G
Only null left

Chemical Identifiers

CAS 2379-60-4
Molecular Formula C8H3Cl2N3O2
Molecular Weight (g/mol) 244.03
MDL Number MFCD00454905
InChI Key SFJCUOAQTGDBPO-UHFFFAOYSA-N
PubChem CID 689090
IUPAC Name 2,3-dichloro-6-nitroquinoxaline
SMILES [O-][N+](=O)C1=CC=C2N=C(Cl)C(Cl)=NC2=C1

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Formula Weight 244.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-6-nitroquinoxaline