missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone 97.0+%, TCI America™

Catalog No. D24211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24211G 1 g
D24215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24211G Supplier TCI America Supplier No. D24211G
Only null left

Chemical Identifiers

CAS 14918-69-5
Molecular Formula C10H4Cl2O4
Molecular Weight (g/mol) 259.038
MDL Number MFCD00075261
InChI Key KFAVXUQFBGHURZ-UHFFFAOYSA-N
PubChem CID 4407
IUPAC Name 6,7-dichloro-5,8-dihydroxynaphthalene-1,4-dione
SMILES C1=CC(=O)C2=C(C1=O)C(=C(C(=C2O)Cl)Cl)O

Specifications

Melting Point 201°C
Quantity 1 g
Formula Weight 259.04
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone