missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone 97.0+%, TCI America™

Supplier:  TCI America D1070250G

 View more versions of this product

Catalog No. D1070250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
214°C
C8Cl2N2O2
250 g
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N
227
227.00
Crystalline Powder
84-58-2
Brown-Yellow
MFCD00001593
3439
HZNVUJQVZSTENZ-UHFFFAOYSA-N
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
6775
≥97.0% (T)

Chemical Identifiers

84-58-2
227
HZNVUJQVZSTENZ-UHFFFAOYSA-N
6775
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N
C8Cl2N2O2
MFCD00001593
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1822

RTECSNumber : GU4825000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions