missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone 97.0+%, TCI America™

Catalog No. p-7126580
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
D107025G 25 g
D1070250G 250 g
2 options

Catalog No. D107025G

Supplier: TCI America D107025G

Only null left

Chemical Identifiers

CAS 84-58-2
Molecular Formula C8Cl2N2O2
Molecular Weight (g/mol) 227
MDL Number MFCD00001593
InChI Key HZNVUJQVZSTENZ-UHFFFAOYSA-N
Synonym 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
PubChem CID 6775
IUPAC Name 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
SMILES C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N

Specifications

Melting Point 214°C
Color Brown-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 227.00
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
EINECSNumber (3)-1822
RTECSNumber GU4825000
TSCA Yes
Recommended Storage Refrigerator