Learn More
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%
CAS: 84-58-2 | C8Cl2N2O2 | 227.01 g/mol
Supplier: Thermo Scientific Chemicals 113301000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
| 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone | |
| 97.5 | |
| 210.0°C to 217.0°C | |
| 0.5% max. (1 g, 80°C, 2 hrs, 40mm Hg vacuum) | |
| 97.5% min. (Iodometry) | |
| C8Cl2N2O2 | |
| 10,902 | |
| 15,3072 | |
| Solubility in water: reacts. Other solubilities: soluble in benzene,dioxane,acetic acid,ethanol,slightly soluble in chloroform,methylenechloride | |
| C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N | |
| 227.01 | |
| 227.01 | |
| Crystalline Powder or Crystals |
| 84-58-2 | |
| 100.0 | |
| Yellow to Orange | |
| Authentic | |
| Glass bottle | |
| 100 g | |
| 01,215; 02,112; 03,83; 04,130; 05,193; 06,168; 07,96; 08,153; 09,148; 10,135; 11,166; 13,104; 14,126; 15,125; 16,120 | |
| 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile | |
| HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
| 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile | |
| 6775 | |
| 98% |
Chemical Identifiers
| 84-58-2 | |
| 227.01 | |
| 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile | |
| 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile |
| C8Cl2N2O2 | |
| HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
| 6775 | |
| C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective glo
GHS Signal Word: Danger
EINECSNumber : 201-542-2
RUO – Research Use Only