missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%

Catalog No. p-4258938
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113300100 10 g Glass bottle
AC113301000 100 g Glass bottle
AC113305000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC113300100 Supplier Thermo Scientific Chemicals Supplier No. 113300100
Only null left

CAS: 84-58-2 | C8Cl2N2O2 | 227.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 84-58-2
Molecular Formula C8Cl2N2O2
Molecular Weight (g/mol) 227.01
InChI Key HZNVUJQVZSTENZ-UHFFFAOYSA-N
Synonym 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
PubChem CID 6775
IUPAC Name 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
SMILES C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 210.0°C to 217.0°C
Color Yellow to Orange
Loss on Drying 0.5% max. (1 g, 80°C, 2 hrs, 40mm Hg vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 10 g
Beilstein 10,902
Fieser 01,215; 02,112; 03,83; 04,130; 05,193; 06,168; 07,96; 08,153; 09,148; 10,135; 11,166; 13,104; 14,126; 15,125; 16,120
Merck Index 15,3072
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene,dioxane,acetic acid,ethanol,slightly soluble in chloroform,methylenechloride
Formula Weight 227.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective glo
EINECSNumber 201-542-2

RUO – Research Use Only