missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopropyl Acrylate 85.0+%, TCI America™

Catalog No. A091025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A091025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A091025G Supplier TCI America Supplier No. A091025G

Chemical Identifiers

CAS 19660-16-3
Molecular Formula C6H8Br2O2
Molecular Weight (g/mol) 271.936
MDL Number MFCD00059461
InChI Key MUKJDVAYJDKPAG-UHFFFAOYSA-N
Synonym Acrylic Acid 2,3-Dibromopropyl Ester
PubChem CID 29728
IUPAC Name 2,3-dibromopropyl prop-2-enoate
SMILES C=CC(=O)OCC(CBr)Br

Specifications

Color Yellow
Boiling Point 105°C
Quantity 25 g
Formula Weight 271.94
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dibromopropyl Acrylate
EINECSNumber (2)-0996
RTECSNumber AS7525000
TSCA Yes