accessibility menu, dialog, popup

UserName

BrBrOOCH2

CAS RN 19660-16-3

IUPAC Name: 2,3-dibromopropyl prop-2-enoate
Synonyms: 2,3-dibromopropyl prop-2-enoate
Molecular Weight (g/mol): 271.94
Molecular Formula: C6H8Br2O2
InChi Key: MUKJDVAYJDKPAG-UHFFFAOYNA-N
SMILES: BrCC(Br)COC(=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results