missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopropionyl Chloride 98.0+%, TCI America™

Supplier:  TCI America D32885G

 View more versions of this product

Catalog No. D32885G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3-Dibromopropionyl Chloride
Yellow
C3H3Br2ClO
5 g
2,3-dibromopropionyl chloride, propanoyl chloride, 2,3-dibromo, alpha,beta-dibromopropionyl chloride, propanoyl chloride,2,3-dibromo, propionyl chloride, 2,3-dibromo, acmc-209eqo, propanoyl chloride, 2,3-dibromo-9ci, 2,3-dibromopropanyl chloride, 3-02-00-00572 beilstein handbook reference
ClC(=O)C(Br)CBr
250.31
250.31
Liquid
18791-02-1
193°C
MFCD00000712
3265
HWKWYDXHMQQDQJ-UHFFFAOYNA-N
2,3-dibromopropanoyl chloride
86790
≥98.0% (T)

Chemical Identifiers

18791-02-1
250.31
HWKWYDXHMQQDQJ-UHFFFAOYNA-N
86790
ClC(=O)C(Br)CBr
C3H3Br2ClO
MFCD00000712
2,3-dibromopropionyl chloride, propanoyl chloride, 2,3-dibromo, alpha,beta-dibromopropionyl chloride, propanoyl chloride,2,3-dibromo, propionyl chloride, 2,3-dibromo, acmc-209eqo, propanoyl chloride, 2,3-dibromo-9ci, 2,3-dibromopropanyl chloride, 3-02-00-00572 beilstein handbook reference
2,3-dibromopropanoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UG6657500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions