missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopropionyl Chloride 98.0+%, TCI America™

Catalog No. p-7127649
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32885G 5 g
D328825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32885G Supplier TCI America Supplier No. D32885G

Chemical Identifiers

CAS 18791-02-1
Molecular Formula C3H3Br2ClO
Molecular Weight (g/mol) 250.31
MDL Number MFCD00000712
InChI Key HWKWYDXHMQQDQJ-UHFFFAOYNA-N
Synonym 2,3-dibromopropionyl chloride, propanoyl chloride, 2,3-dibromo, alpha,beta-dibromopropionyl chloride, propanoyl chloride,2,3-dibromo, propionyl chloride, 2,3-dibromo, acmc-209eqo, propanoyl chloride, 2,3-dibromo-9ci, 2,3-dibromopropanyl chloride, 3-02-00-00572 beilstein handbook reference
PubChem CID 86790
IUPAC Name 2,3-dibromopropanoyl chloride
SMILES ClC(=O)C(Br)CBr

Specifications

Color Yellow
Boiling Point 193°C
Quantity 5 g
UN Number 3265
Formula Weight 250.31
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2,3-Dibromopropionyl Chloride
RTECSNumber UG6657500
TSCA No