missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dibromopropionyl Chloride 98.0+%, TCI America™
Supplier: TCI America D328825G
Specifications
| 2,3-Dibromopropionyl Chloride | |
| Yellow | |
| C3H3Br2ClO | |
| 25 g | |
| 2,3-dibromopropionyl chloride, propanoyl chloride, 2,3-dibromo, alpha,beta-dibromopropionyl chloride, propanoyl chloride,2,3-dibromo, propionyl chloride, 2,3-dibromo, acmc-209eqo, propanoyl chloride, 2,3-dibromo-9ci, 2,3-dibromopropanyl chloride, 3-02-00-00572 beilstein handbook reference | |
| ClC(=O)C(Br)CBr | |
| 250.31 | |
| 250.31 | |
| Liquid |
| 18791-02-1 | |
| 193°C | |
| MFCD00000712 | |
| 3265 | |
| HWKWYDXHMQQDQJ-UHFFFAOYNA-N | |
| 2,3-dibromopropanoyl chloride | |
| 86790 | |
| ≥98.0% (T) |
Chemical Identifiers
| 18791-02-1 | |
| 250.31 | |
| HWKWYDXHMQQDQJ-UHFFFAOYNA-N | |
| 86790 | |
| ClC(=O)C(Br)CBr |
| C3H3Br2ClO | |
| MFCD00000712 | |
| 2,3-dibromopropionyl chloride, propanoyl chloride, 2,3-dibromo, alpha,beta-dibromopropionyl chloride, propanoyl chloride,2,3-dibromo, propionyl chloride, 2,3-dibromo, acmc-209eqo, propanoyl chloride, 2,3-dibromo-9ci, 2,3-dibromopropanyl chloride, 3-02-00-00572 beilstein handbook reference | |
| 2,3-dibromopropanoyl chloride |
Safety and Handling
RTECSNumber : UG6657500
TSCA : No