missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopropionic Acid 97.0+%, TCI America™

Catalog No. p-7126702
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D138325G 25 g
D1383500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D138325G Supplier TCI America Supplier No. D138325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 600-05-5
Molecular Formula C3H4Br2O2
Molecular Weight (g/mol) 231.87
MDL Number MFCD00004212
InChI Key ZMYAKSMZTVWUJB-UHFFFAOYNA-N
Synonym 2,3-dibromopropionic acid, propanoic acid, 2,3-dibromo, alpha,beta-dibromopropionic acid, propionic acid, 2,3-dibromo, .alpha.,.beta.-dibromopropionic acid, acmc-209mhg, propanoic acid,3-dibromo, propionic acid,3-dibromo, 2,3-dibromo-propionic acid, dsstox_cid_12832
PubChem CID 11746
IUPAC Name 2,3-dibromopropanoic acid
SMILES OC(=O)C(Br)CBr

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 119°C
Quantity 25 g
UN Number 3261
Formula Weight 231.87
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromopropionic Acid
RTECSNumber UF0660000
TSCA Yes
Recommended Storage Refrigerator