missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dibromonaphthalene 98.0+%, TCI America™
$78.29 - $233.00
Chemical Identifiers
| CAS | 13214-70-5 |
|---|---|
| Molecular Formula | C10H6Br2 |
| Molecular Weight (g/mol) | 285.97 |
| MDL Number | MFCD00021589 |
| InChI Key | GTILXPRQNNYDHT-UHFFFAOYSA-N |
| Synonym | naphthalene, 2,3-dibromo, 2,3-dibromo-naphthalene, acmc-209uxy, pbn-10 |
| PubChem CID | 123298 |
| IUPAC Name | 2,3-dibromonaphthalene |
| SMILES | BrC1=C(Br)C=C2C=CC=CC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4597200MG
|
TCI America
D4597200MG |
200 mg |
Each for $78.29
|
|
|||||
|
D45971G
|
TCI America
D45971G |
1 g |
Each for $233.00
|
|
|||||
Chemical Identifiers
| 13214-70-5 | |
| 285.97 | |
| GTILXPRQNNYDHT-UHFFFAOYSA-N | |
| 123298 | |
| BrC1=C(Br)C=C2C=CC=CC2=C1 |
| C10H6Br2 | |
| MFCD00021589 | |
| naphthalene, 2,3-dibromo, 2,3-dibromo-naphthalene, acmc-209uxy, pbn-10 | |
| 2,3-dibromonaphthalene |
Specifications
| 13214-70-5 | |
| White-Yellow | |
| MFCD00021589 | |
| naphthalene, 2,3-dibromo, 2,3-dibromo-naphthalene, acmc-209uxy, pbn-10 | |
| BrC1=C(Br)C=C2C=CC=CC2=C1 | |
| 285.97 | |
| 285.97 | |
| Crystalline Powder |
| 142°C | |
| C10H6Br2 | |
| 200 mg | |
| GTILXPRQNNYDHT-UHFFFAOYSA-N | |
| 2,3-dibromonaphthalene | |
| 123298 | |
| ≥98.0% (GC) | |
| 2,3-Dibromonaphthalene |
Safety and Handling
TSCA : No