accessibility menu, dialog, popup

UserName

CAS RN 13214-70-5

BrBr

CAS RN 13214-70-5

IUPAC Name: 2,3-dibromonaphthalene
Synonyms: 2,3-dibromonaphthalene
Molecular Weight (g/mol): 285.97
Molecular Formula: C10H6Br2
InChi Key: GTILXPRQNNYDHT-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results