missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromofuran, 97%, stab. with 0.5% calcium carbonate

Catalog No. p-7046645
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1551803 1 g
AAL1551806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1551803 Supplier Thermo Scientific Chemicals Supplier No. L1551803
Only null left

CAS: 30544-34-4 | C4H2Br2O | 225.867 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30544-34-4
Molecular Formula C4H2Br2O
Molecular Weight (g/mol) 225.867
MDL Number MFCD01074839
InChI Key GKPGEBCMRMQOPF-UHFFFAOYSA-N
Synonym 2,3-dibrom-furan, acmc-1adxx, furan, 2,3-dibromo, 2,3-bis bromanyl furan
PubChem CID 7021501
IUPAC Name 2,3-dibromofuran
SMILES C1=COC(=C1Br)Br

Specifications

Density 2
Boiling Point 75°C to 78°C (75 mmHg)
Refractive Index 1.547
Quantity 1 g
Beilstein 107511
Sensitivity Light sensitive
Formula Weight 225.87
Percent Purity 97%
Chemical Name or Material 2,3-Dibromofuran, stabilized with 0.5% calcium carbonate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only