accessibility menu, dialog, popup

UserName

OBrBr

CAS RN 30544-34-4

IUPAC Name: 2,3-dibromofuran
Synonyms: 2,3-dibromofuran
Molecular Weight (g/mol): 225.87
Molecular Formula: C4H2Br2O
InChi Key: GKPGEBCMRMQOPF-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)