missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde, 98%

CAS: 2973-75-3 | C8H6Br2O3 | 309.94 g/mol

$55.08 - $630.22

Chemical Identifiers

CAS 2973-75-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00016978
InChI Key WKLKGSHBXNPUDU-UHFFFAOYSA-N
Synonym 5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
PubChem CID 520452
IUPAC Name 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=C(Br)C(Br)=C1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2114003
View Documents
Thermo Scientific Chemicals
B2114003
1 g
Each for $55.08
Only null left
 
AAB2114006
View Documents
Thermo Scientific Chemicals
B2114006
5 g
Each for $163.82
Only null left
 
AAB2114014
View Documents
Thermo Scientific Chemicals
B2114014
25 g
Each for $630.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2973-75-3
309.94
WKLKGSHBXNPUDU-UHFFFAOYSA-N
520452
COC1=CC(C=O)=C(Br)C(Br)=C1O
C8H6Br2O3
MFCD00016978
5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde

Specifications

2973-75-3
C8H6Br2O3
1 g
Air Sensitive
WKLKGSHBXNPUDU-UHFFFAOYSA-N
2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
520452
98%
213°C to 217°C
MFCD00016978
1968576
5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
COC1=CC(C=O)=C(Br)C(Br)=C1O
309.94
309.94
2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only