missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diaminotoluene 97.0+%, TCI America™
$329.94 - $1,219.10
Chemical Identifiers
| CAS | 2687-25-4 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.17 |
| MDL Number | MFCD00011589 |
| InChI Key | AXNUJYHFQHQZBE-UHFFFAOYSA-N |
| Synonym | 2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine |
| PubChem CID | 17593 |
| IUPAC Name | 3-methylbenzene-1,2-diamine |
| SMILES | CC1=CC=CC(N)=C1N |
Chemical Identifiers
| 2687-25-4 | |
| 122.17 | |
| AXNUJYHFQHQZBE-UHFFFAOYSA-N | |
| 17593 | |
| CC1=CC=CC(N)=C1N |
| C7H10N2 | |
| MFCD00011589 | |
| 2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine | |
| 3-methylbenzene-1,2-diamine |
Specifications
| 2687-25-4 | |
| 225°C | |
| MFCD00011589 | |
| 2811 | |
| AXNUJYHFQHQZBE-UHFFFAOYSA-N | |
| 3-methylbenzene-1,2-diamine | |
| 17593 | |
| ≥97.0% (GC,T) | |
| 2,3-Diaminotoluene |
| 60°C | |
| C7H10N2 | |
| 5 g | |
| 2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine | |
| CC1=CC=CC(N)=C1N | |
| 122.17 | |
| 122.17 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0126
RTECSNumber : XS9550000
TSCA : Yes