missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminotoluene 97.0+%, TCI America™

Supplier:  TCI America D37885G

 View more versions of this product

Catalog No. D37885G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3-Diaminotoluene
60°C
C7H10N2
5 g
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
CC1=CC=CC(N)=C1N
122.17
122.17
Crystalline Powder
2687-25-4
225°C
MFCD00011589
2811
AXNUJYHFQHQZBE-UHFFFAOYSA-N
3-methylbenzene-1,2-diamine
17593
≥97.0% (GC,T)

Chemical Identifiers

2687-25-4
122.17
AXNUJYHFQHQZBE-UHFFFAOYSA-N
17593
CC1=CC=CC(N)=C1N
C7H10N2
MFCD00011589
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
3-methylbenzene-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0126

RTECSNumber : XS9550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions