missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminopyridine 98.0+%, TCI America™

Catalog No. p-7126045
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D01175G 5 g
D011725G 25 g
2 options

Catalog No. D01175G

Supplier: TCI America D01175G

Only null left

Chemical Identifiers

CAS 452-58-4
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006319
InChI Key ZZYXNRREDYWPLN-UHFFFAOYSA-N
Synonym 2,3-diaminopyridine, 2,3-pyridinediamine, pyridine, 2,3-diamino, pyridine-2,3-diyldiamine, unii-cbx394737h, diaminopyridine, 2,3-diamino-pyridine, pyridinediamine, pyridine,3-diamino, pubchem1296
PubChem CID 9956
IUPAC Name pyridine-2,3-diamine
SMILES NC1=CC=CN=C1N

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 109.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diaminopyridine
EINECSNumber (5)-3702
RTECSNumber US7548000
TSCA No