accessibility menu, dialog, popup

UserName

NH2NH2N

CAS RN 452-58-4

IUPAC Name: pyridine-2,3-diamine
Synonyms: pyridine-2,3-diamine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: ZZYXNRREDYWPLN-UHFFFAOYSA-N
SMILES: NC1=CC=CN=C1N
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)