missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminopyridine, 98%

CAS: 452-58-4 | C5H7N3 | 109.13 g/mol

Supplier:  Thermo Scientific Chemicals A1461814

 View more versions of this product

Catalog No. AAA1461814


Only null left
Explore available promotions

Description

Description

2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

Solubility
Soluble in water (100 g/L).

Notes
It is sensitive to air. Incompatible with oxidizing agents.
Specifications

Specifications

2,3-Diaminopyridine
110°C to 114°C
205°C (401°F)
MFCD00006319
UN2671
Air Sensitive
Soluble in water (100g/L).
NC1=CC=CN=C1N
109.13
109.13
452-58-4
195°C (55 mmHg)
C5H7N3
25 g
109869
2,3-diaminopyridine, 2,3-pyridinediamine, pyridine, 2,3-diamino, pyridine-2,3-diyldiamine, unii-cbx394737h, diaminopyridine, 2,3-diamino-pyridine, pyridinediamine, pyridine,3-diamino, pubchem1296
ZZYXNRREDYWPLN-UHFFFAOYSA-N
pyridine-2,3-diamine
9956
98%

Chemical Identifiers

452-58-4
109.13
ZZYXNRREDYWPLN-UHFFFAOYSA-N
9956
NC1=CC=CN=C1N
C5H7N3
MFCD00006319
2,3-diaminopyridine, 2,3-pyridinediamine, pyridine, 2,3-diamino, pyridine-2,3-diyldiamine, unii-cbx394737h, diaminopyridine, 2,3-diamino-pyridine, pyridinediamine, pyridine,3-diamino, pubchem1296
pyridine-2,3-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES

EINECSNumber : 207-200-9

RTECSNumber : US7548000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only