missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diaminopyridine, 98%
Description
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
Solubility
Soluble in water (100 g/L).
Notes
It is sensitive to air. Incompatible with oxidizing agents.
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
Solubility
Soluble in water (100 g/L).
Notes
It is sensitive to air. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 110°C to 114°C |
| Boiling Point | 195°C (55 mmHg) |
| Flash Point | 205°C (401°F) |
| Quantity | 100 g |
| UN Number | UN2671 |
| Beilstein | 109869 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in water (100g/L). |
| Formula Weight | 109.13 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H301-H312+H332-H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H300-H311-H331-H315-H319-H335 Fatal if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES |
| EINECSNumber | 207-200-9 |
| RTECSNumber | US7548000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only