missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminopyridine, 98%

Catalog No. AAA1461806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1461806 5 g
AAA1461814 25 g
AAA1461822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1461806 Supplier Thermo Scientific Chemicals Supplier No. A1461806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 452-58-4 | C5H7N3 | 109.13 g/mol

2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

Solubility
Soluble in water (100 g/L).

Notes
It is sensitive to air. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 452-58-4
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006319
InChI Key ZZYXNRREDYWPLN-UHFFFAOYSA-N
Synonym 2,3-diaminopyridine, 2,3-pyridinediamine, pyridine, 2,3-diamino, pyridine-2,3-diyldiamine, unii-cbx394737h, diaminopyridine, 2,3-diamino-pyridine, pyridinediamine, pyridine,3-diamino, pubchem1296
PubChem CID 9956
IUPAC Name pyridine-2,3-diamine
SMILES NC1=CC=CN=C1N

Specifications

Melting Point 110°C to 114°C
Boiling Point 195°C (55 mmHg)
Flash Point 205°C (401°F)
Quantity 5 g
UN Number UN2671
Beilstein 109869
Sensitivity Air Sensitive
Solubility Information Soluble in water (100g/L).
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2,3-Diaminopyridine
Show More Show Less
Hazard Category H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
EINECSNumber 207-200-9
RTECSNumber US7548000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only