missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminophenol, 97%

Catalog No. AC425680010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425680010 1 g Glass bottle
AC425680050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425680010 Supplier Thermo Scientific Chemicals Supplier No. 425680010

CAS: 59649-56-8 | C6H8N2O | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59649-56-8
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD00075199
InChI Key PCAXITAPTVOLGL-UHFFFAOYSA-N
Synonym diaminophenol, phenol, 2,3-diamino, phenol, diamino, zlchem 936, 2,3-diamino-phenol, pubchem2129, acmc-209mex, 2,3-bis azanyl phenol, 2,3-diaminophenol, 3-hydroxy-o-phenylenediamine
PubChem CID 579937
IUPAC Name 2,3-diaminophenol
SMILES C1=CC(=C(C(=C1)O)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 161°C to 165°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (NH2)2C6H3OH
Quantity 1 g
Solubility Information Solubility in water: not soluble
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Granules
Chemical Name or Material 2, 3-Diaminophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only