missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminonaphthalene 98.0+%, TCI America™

Catalog No. D4101100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4101100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4101100MG Supplier TCI America Supplier No. D4101100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 771-97-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00004116
InChI Key XTBLDMQMUSHDEN-UHFFFAOYSA-N
Synonym 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532
PubChem CID 69872
IUPAC Name naphthalene-2,3-diamine
SMILES C1=CC=C2C=C(C(=CC2=C1)N)N

Specifications

Melting Point 198°C
Quantity 100 mg
Formula Weight 158.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diaminonaphthalene [for Biochemical Research]
TSCA Yes
Recommended Storage Refrigerator