missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diamino-5-bromopyridine 98.0+%, TCI America™
$50.33 - $158.44
Chemical Identifiers
| CAS | 38875-53-5 |
|---|---|
| Molecular Formula | C5H7BrN3 |
| Molecular Weight (g/mol) | 189.04 |
| MDL Number | MFCD00460094 |
| InChI Key | YRGMYJUKFJPNPD-UHFFFAOYSA-O |
| Synonym | 2,3-diamino-5-bromopyridine, 5-bromo-2,3-diaminopyridine, 5-bromo-2,3-pyridinediamine, 2,3-pyridinediamine, 5-bromo, 3-amino-5-bromopyridin-2-yl amine, abbypharma ap-17-5972, 5-brompyridin-2,3-diamin, pubchem5383, acmc-1adjl, 5-brompyridin-2,3-diamine |
| PubChem CID | 691156 |
| IUPAC Name | 2,3-diamino-5-bromopyridin-1-ium |
| SMILES | NC1=CC(Br)=C[NH+]=C1N |
Chemical Identifiers
| 38875-53-5 | |
| 189.04 | |
| YRGMYJUKFJPNPD-UHFFFAOYSA-O | |
| 691156 | |
| NC1=CC(Br)=C[NH+]=C1N |
| C5H7BrN3 | |
| MFCD00460094 | |
| 2,3-diamino-5-bromopyridine, 5-bromo-2,3-diaminopyridine, 5-bromo-2,3-pyridinediamine, 2,3-pyridinediamine, 5-bromo, 3-amino-5-bromopyridin-2-yl amine, abbypharma ap-17-5972, 5-brompyridin-2,3-diamin, pubchem5383, acmc-1adjl, 5-brompyridin-2,3-diamine | |
| 2,3-diamino-5-bromopyridin-1-ium |
Specifications
| 38875-53-5 | |
| C5H7BrN3 | |
| 5 g | |
| YRGMYJUKFJPNPD-UHFFFAOYSA-O | |
| 2,3-diamino-5-bromopyridin-1-ium | |
| 691156 | |
| ≥98.0% (GC,T) | |
| 2,3-Diamino-5-bromopyridine |
| 161°C | |
| MFCD00460094 | |
| 2,3-diamino-5-bromopyridine, 5-bromo-2,3-diaminopyridine, 5-bromo-2,3-pyridinediamine, 2,3-pyridinediamine, 5-bromo, 3-amino-5-bromopyridin-2-yl amine, abbypharma ap-17-5972, 5-brompyridin-2,3-diamin, pubchem5383, acmc-1adjl, 5-brompyridin-2,3-diamine | |
| NC1=CC(Br)=C[NH+]=C1N | |
| 189.04 | |
| 188.03 | |
| Crystalline Powder |
Safety and Handling
TSCA : No